Tisk…

Methyl-3-amino-5-fluorobenzoate 98%

Dodavatel: Apollo Scientific

PC201353-25G PC201353-5G
APOSPC201353-25GEA 11310 CZK
APOSPC201353-25G APOSPC201353-5G
Methyl-3-amino-5-fluorobenzoate 98%
Methyl-3-amino-5-fluorobenzoate
Vzorec: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD05863683
CAS číslo: 884497-46-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR