Tisk…

(3-Fluoropyridin-4-yl)methanamine 96%

Dodavatel: Apollo Scientific

PC201335-1G PC201335-250MG PC201335-5G
APOSPC201335-1GEA 4140 CZK
APOSPC201335-1G APOSPC201335-250MG APOSPC201335-5G
(3-Fluoropyridin-4-yl)methanamine 96%
(3-Fluoropyridin-4-yl)methanamine
Vzorec: C₆H₇FN₂
Storage Temperature: Refrigerator
MDL Number: MFCD11100727
CAS číslo: 870063-62-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR