Tisk…

(R)-1-(4-(Trifluoromethyl)phenyl)ethanamine hydrochloride 95+%

Dodavatel: Apollo Scientific

PC201325-1G PC201325-250MG PC201325-5G
APOSPC201325-1GEA 9660 CZK
APOSPC201325-1G APOSPC201325-250MG APOSPC201325-5G
(R)-1-(4-(Trifluoromethyl)phenyl)ethanamine hydrochloride 95+%
(R)-1-(4-(Trifluoromethyl)phenyl)ethanamine hydrochloride
Vzorec: C₉H₁₀F₃N·ClH
Storage Temperature: Ambient
MDL Number: MFCD11100948
CAS číslo: 856645-99-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR