Tisk…

5-Chloro-4-(trifluoromethyl)pyridine-2-carboxylic acid 96%

Dodavatel: Apollo Scientific
Varování

PC201310-250MG
APOSPC201310-250MGEA 16530 CZK
APOSPC201310-250MG
5-Chloro-4-(trifluoromethyl)pyridine-2-carboxylic acid 96%
5-Chloro-4-(trifluoromethyl)pyridine-2-carboxylic acid
Vzorec: C₇H₃ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD18256834
CAS číslo: 796090-31-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR