Tisk…

Methyl 3-fluoro-2-hydroxybenzoate 98%

Dodavatel: Apollo Scientific
Varování

PC201284-250MG PC201284-1G PC201284-5G
APOSPC201284-250MGEA 1660 CZK
APOSPC201284-250MG APOSPC201284-1G APOSPC201284-5G
Methyl 3-fluoro-2-hydroxybenzoate 98%
Methyl 3-fluoro-2-hydroxybenzoate
Vzorec: C₈H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD06203651
CAS číslo: 70163-98-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR