Tisk…

Methyl-4-fluoro-3-(trifluoromethyl)benzoate 97+%

Dodavatel: Apollo Scientific

Synonyms: Methyl 4-fluoro-3-(trifluoromethyl)benzoate

PC201175-10G
APOSPC201175-10GEA 2570 CZK
APOSPC201175-10G
Methyl-4-fluoro-3-(trifluoromethyl)benzoate 97+%
Methyl-4-fluoro-3-(trifluoromethyl)benzoate
Vzorec: C₉H₆F₄O₂
MW: 222.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203709
CAS číslo: 176694-36-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR