Tisk…

2-Amino-4,6-difluorobenzaldehyde 98%

Dodavatel: Apollo Scientific

PC201099-100MG PC201099-1G PC201099-250MG
APOSPC201099-100MGEA 4400 CZK
APOSPC201099-100MG APOSPC201099-1G APOSPC201099-250MG
2-Amino-4,6-difluorobenzaldehyde 98%
2-Amino-4,6-difluorobenzaldehyde
Vzorec: C₇H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD15528759
CAS číslo: 1260790-53-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR