Tisk…

4-Chloro-8-fluoro-2-methylquinoline

Dodavatel: Apollo Scientific

PC200565-1G PC200565-5G
APOSPC200565-1GEA 5720 CZK
APOSPC200565-1G APOSPC200565-5G
4-Chloro-8-fluoro-2-methylquinoline
4-Chloro-8-fluoro-2-methylquinoline
Vzorec: C₁₀H₇ClFN
Storage Temperature: Ambient
MDL Number: MFCD00272335
CAS číslo: 18615-59-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR