Tisk…

(RS)-1-(3,5-Difluorophenyl)ethylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC2002-1G PC2002-25G PC2002-5G
APOSPC2002-1GEA 744 CZK
APOSPC2002-1G APOSPC2002-25G APOSPC2002-5G
(RS)-1-(3,5-Difluorophenyl)ethylamine 97%
(RS)-1-(3,5-Difluorophenyl)ethylamine
Vzorec: C₈H₉F₂N
Bod varu: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD04038286
CAS číslo: 321318-29-0
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR