Tisk…

Potassium (1-naphthalene)trifluoroborate

Dodavatel: Apollo Scientific

PC200179-10G PC200179-1G PC200179-5G
APOSPC200179-10GEA 7280 CZK
APOSPC200179-10G APOSPC200179-1G APOSPC200179-5G
Potassium (1-naphthalene)trifluoroborate
Potassium (1-naphthalene)trifluoroborate
Vzorec: C₁₀H₇BF₃K
Storage Temperature: Ambient
MDL Number: MFCD06659936
CAS číslo: 166328-07-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR