Tisk…

Potassium 4-((morpholino)methyl) phenyltrifluoroborate

Dodavatel: Apollo Scientific

PC200158-1G PC200158-500MG PC200158-5G
APOSPC200158-1GEA 3640 CZK
APOSPC200158-1G APOSPC200158-500MG APOSPC200158-5G
Potassium 4-((morpholino)methyl) phenyltrifluoroborate
Potassium 4-((morpholino)methyl) phenyltrifluoroborate
Vzorec: C₁₁H₁₄BF₃NO·K
Storage Temperature: Ambient
MDL Number: MFCD20527095
CAS číslo: 1190095-06-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR