Tisk…

Potassium-3-carboxyphenyltrifluoroborate

Dodavatel: Apollo Scientific

PC200140-10G PC200140-1G PC200140-5G
APOSPC200140-10GEA 9690 CZK
APOSPC200140-10G APOSPC200140-1G APOSPC200140-5G
Potassium-3-carboxyphenyltrifluoroborate
Potassium-3-carboxyphenyltrifluoroborate
Vzorec: C₇H₅BF₃KO₂
Storage Temperature: Ambient
MDL Number: MFCD07784423
CAS číslo: 850313-91-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR