Tisk…

2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol

Dodavatel: Apollo Scientific

PC200130-1G PC200130-500MG PC200130-5G
APOSPC200130-1GEA 3340 CZK
APOSPC200130-1G APOSPC200130-500MG APOSPC200130-5G
2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol
2-[(4-Fluorophenyl)dimethylsilyl]benzyl alcohol
Vzorec: C₁₅H₁₇FOSi
Storage Temperature: Ambient
MDL Number: MFCD16251550
CAS číslo: 853955-70-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR