Tisk…

Potassium-2-(ethoxycarbonyl)phenyltrifluoroborate 98%

Dodavatel: Apollo Scientific

PC1768-5G
APOSPC1768-5GEA 2240 CZK
APOSPC1768-5G
Potassium-2-(ethoxycarbonyl)phenyltrifluoroborate 98%
Potassium-2-(ethoxycarbonyl)phenyltrifluoroborate
Vzorec: C₉H₉BF₃KO₂
Storage Temperature: Ambient
MDL Number: MFCD04115768
CAS číslo: 850623-73-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR