Tisk…

Perfluoro-3,6,9-trioxatridecanoic acid 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC1737-100G PC1737-25G PC1737-1G PC1737-5G
APOSPC1737-100GEA 0 CZK
APOSPC1737-100G APOSPC1737-25G APOSPC1737-1G APOSPC1737-5G
Perfluoro-3,6,9-trioxatridecanoic acid 98%
Perfluoro-3,6,9-trioxatridecanoic acid
Vzorec: C₁₀HF₁₉O₅
Bod varu: 75 °C/0,8mmHg
Hustota: 1,8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094135
CAS číslo: 330562-41-9
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR