Tisk…

Potassium-2-(hydroxymethyl)phenyltrifluoroborate 96%

Dodavatel: Apollo Scientific

PC1723-100G PC1723-25G PC1723-5G
APOSPC1723-100GEA 42320 CZK
APOSPC1723-100G APOSPC1723-25G APOSPC1723-5G
Potassium-2-(hydroxymethyl)phenyltrifluoroborate 96%
Potassium-2-(hydroxymethyl)phenyltrifluoroborate
Vzorec: C₇H₇BF₃KO
Bod tání: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115769
CAS číslo: 850623-74-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR