Tisk…

3-(Difluoromethoxy)benzeneboronic acid 99%

Dodavatel: Apollo Scientific
Varování

PC1596-1G PC1596-10G PC1596-5G PC1596-25G
APOSPC1596-1GEA 2440 CZK
APOSPC1596-1G APOSPC1596-10G APOSPC1596-5G APOSPC1596-25G
3-(Difluoromethoxy)benzeneboronic acid 99%
3-(Difluoromethoxy)benzeneboronic acid
Vzorec: C₇H₇BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08706282
CAS číslo: 866607-09-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR