Tisk…

3,6-Difluoro-2-methoxybenzaldehyde 98%

Dodavatel: Apollo Scientific

PC1143-1G PC1143-5G PC1143-25G
APOSPC1143-1GEA 870 CZK
APOSPC1143-1G APOSPC1143-5G APOSPC1143-25G
3,6-Difluoro-2-methoxybenzaldehyde 98%
3,6-Difluoro-2-methoxybenzaldehyde
Vzorec: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD08275289
CAS číslo: 887267-04-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR