Tisk…

4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde

Dodavatel: Apollo Scientific
Varování

PC11174-1G PC11174-250MG
APOSPC11174-1GEA 910 CZK
APOSPC11174-1G APOSPC11174-250MG
4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde
4'-(Trifluoromethyl)[1,1'-biphenyl]-3-carbaldehyde
Vzorec: C₁₄H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD01631879
CAS číslo: 343604-24-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR