Tisk…

Difluoromethyl phenyl sulphide 98%

Dodavatel: Apollo Scientific

PC10684-1G PC10684-5G
APOSPC10684-1GEA 2610 CZK
APOSPC10684-1G APOSPC10684-5G
Difluoromethyl phenyl sulphide 98%
Difluoromethyl phenyl sulphide
Vzorec: C₇H₆F₂S
Storage Temperature: Ambient
MDL Number: MFCD16301013
CAS číslo: 1535-67-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR