Tisk…

2-Methyl-4-(trifluoromethoxy)benzoyl chloride 95+%

Dodavatel: Apollo Scientific
Nebezpečí

PC1063-1G PC1063-5G
APOSPC1063-1GEA 3740 CZK
APOSPC1063-1G APOSPC1063-5G
2-Methyl-4-(trifluoromethoxy)benzoyl chloride 95+%
2-Methyl-4-(trifluoromethoxy)benzoyl chloride
Vzorec: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD01631544
CAS číslo: 261951-93-3
UN: 1760
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR