Tisk…

Pentafluorophenyl-3-methylbenzenesulphonate

Dodavatel: Apollo Scientific

PC10320-1G PC10320-500MG PC10320-5G
APOSPC10320-1GEA 7840 CZK
APOSPC10320-1G APOSPC10320-500MG APOSPC10320-5G
Pentafluorophenyl-3-methylbenzenesulphonate
Pentafluorophenyl-3-methylbenzenesulphonate
Vzorec: C₁₃H₇F₅O₃S
Bod tání: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD05975125
CAS číslo: 885950-34-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR