Tisk…

Pentafluorophenyl-3-nitrobenzenesulphonate

Dodavatel: Apollo Scientific

PC10313-1G PC10313-500MG PC10313-5G
APOSPC10313-1GEA 7840 CZK
APOSPC10313-1G APOSPC10313-500MG APOSPC10313-5G
Pentafluorophenyl-3-nitrobenzenesulphonate
Pentafluorophenyl-3-nitrobenzenesulphonate
Vzorec: C₁₂H₄F₅NO₅S
Bod tání: 67…68 °C
Storage Temperature: Ambient
MDL Number: MFCD05975121
CAS číslo: 663175-93-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR