Tisk…

Pentafluorophenyl-3-(trifluoromethyl)benzenesulphonate

Dodavatel: Apollo Scientific

PC10310-1G PC10310-500MG PC10310-5G
APOSPC10310-1GEA 7840 CZK
APOSPC10310-1G APOSPC10310-500MG APOSPC10310-5G
Pentafluorophenyl-3-(trifluoromethyl)benzenesulphonate
Pentafluorophenyl-3-(trifluoromethyl)benzenesulphonate
Vzorec: C₁₃H₄F₈O₃S
Storage Temperature: Ambient
MDL Number: MFCD05975128
CAS číslo: 885950-36-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR