Tisk…

1H,1H,8H-Perfluoro-1-octanol

Dodavatel: Apollo Scientific
Varování

PC0935-1G PC0935-5G
APOSPC0935-1GEA 2700 CZK
APOSPC0935-1G APOSPC0935-5G
1H,1H,8H-Perfluoro-1-octanol
1H,1H,8H-Perfluoro-1-octanol
Vzorec: C₈H₄F₁₄O
Storage Temperature: Ambient
MDL Number: MFCD01631687
CAS číslo: 10331-08-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR