Tisk…

3-(4-Fluorophenoxy)benzyl bromide 97%

Dodavatel: Apollo Scientific

PC0853-1G PC0853-5G
APOSPC0853-1GEA 9330 CZK
APOSPC0853-1G APOSPC0853-5G
3-(4-Fluorophenoxy)benzyl bromide 97%
3-(4-Fluorophenoxy)benzyl bromide
Vzorec: C₁₃H₁₀BrFO
Bod varu: 160 °C/2mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00059915
CAS číslo: 65295-58-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR