Tisk…

2-Acetamido-4-fluorobenzoic acid

Dodavatel: Apollo Scientific
Varování

PC0673-100MG PC0673-1G PC0673-5G
APOSPC0673-100MGEA 622 CZK
APOSPC0673-100MG APOSPC0673-1G APOSPC0673-5G
2-Acetamido-4-fluorobenzoic acid
2-Acetamido-4-fluorobenzoic acid
Vzorec: C₉H₈FNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00665891
CAS číslo: 394-27-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR