Tisk…

4-Methyl-3-(trifluoromethyl)benzoyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

PC0469-10G PC0469-1G PC0469-5G
APOSPC0469-10GEA 9430 CZK
APOSPC0469-10G APOSPC0469-1G APOSPC0469-5G
4-Methyl-3-(trifluoromethyl)benzoyl chloride
4-Methyl-3-(trifluoromethyl)benzoyl chloride
Vzorec: C₉H₆ClF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD01631602
CAS číslo: 261952-11-8
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR