Tisk…

3-Amino-5-methylbenzotrifluoride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC0435-1G PC0435-5G
APOSPC0435-1GEA 3000 CZK
APOSPC0435-1G APOSPC0435-5G
3-Amino-5-methylbenzotrifluoride 97%
3-Amino-5-methylbenzotrifluoride
Vzorec: C₈H₈F₃N
Storage Temperature: Ambient
MDL Number: MFCD01631580
CAS číslo: 96100-12-8
UN: 2810
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR