Tisk…

2-Methyl-4-(trifluoromethoxy)aniline 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC0414-25G PC0414-5G PC0414-100G
APOSPC0414-25GEA 5960 CZK
APOSPC0414-25G APOSPC0414-5G APOSPC0414-100G
2-Methyl-4-(trifluoromethoxy)aniline 97%
2-Methyl-4-(trifluoromethoxy)aniline
Vzorec: C₈H₈F₃NO
Storage Temperature: Ambient
MDL Number: MFCD01631541
CAS číslo: 86256-59-9
UN: 2810
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR