Tisk…

4-Fluoro-3-methylbenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Bromomethyl)-1-fluoro-2-methylbenzene

PC0363-10G PC0363-1G PC0363-5G PC0363-25G
APOSPC0363-10GEA 8040 CZK
APOSPC0363-10G APOSPC0363-1G APOSPC0363-5G APOSPC0363-25G
4-Fluoro-3-methylbenzyl bromide
4-Fluoro-3-methylbenzyl bromide
Vzorec: C₈H₈BrF
MW: 203.05
Bod varu: 80 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD01631425
CAS číslo: 261951-70-6
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR