Tisk…

α-Bromo-5-chloro-2-fluorotoluene

Dodavatel: Apollo Scientific
Nebezpečí

PC0080-10G PC0080-5G PC0080-25G
APOSPC0080-10GEA 827 CZK
APOSPC0080-10G APOSPC0080-5G APOSPC0080-25G
α-Bromo-5-chloro-2-fluorotoluene
α-Bromo-5-chloro-2-fluorotoluene
Vzorec: C₇H₅BrClF
Bod varu: 44…45 °C/0,5mmHg
Storage Temperature: Ambient
MDL Number: MFCD01631437
CAS číslo: 71916-91-1
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR