Tisk…

3-Chloro-2,6-difluorobenzoyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Chloro-2,6-difluorobenzene-1-carbonyl chloride

PC0001-1G PC0001-5G
APOSPC0001-1GEA 827 CZK
APOSPC0001-1G APOSPC0001-5G
3-Chloro-2,6-difluorobenzoyl chloride 98%
3-Chloro-2,6-difluorobenzoyl chloride
Vzorec: C₇H₂Cl₂F₂O
MW: 210.99
Bod varu: 222
Bod tání: 70
Storage Temperature: Ambient
MDL Number: MFCD01631324
CAS číslo: 261762-43-0
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR