Tisk…

2-Amino-5-chlorothiophenol 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR9714-1G OR9714-250MG
APOSOR9714-1GEA 16170 CZK
APOSOR9714-1G APOSOR9714-250MG
2-Amino-5-chlorothiophenol 98%
2-Amino-5-chlorothiophenol
Vzorec: C₆H₆ClNS
Storage Temperature: Refrigerator
CAS číslo: 23474-98-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR