Tisk…

2-(Methylsulphonyl)benzenesulphonyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR9710-1G
APOSOR9710-1GEA 1330 CZK
APOSOR9710-1G
2-(Methylsulphonyl)benzenesulphonyl chloride 95%
2-(Methylsulphonyl)benzenesulphonyl chloride
Vzorec: C₇H₇ClO₄S₂
Bod tání: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00216493
CAS číslo: 89265-35-0
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR