Tisk…

Z-D-MEAla-OH 97%

Dodavatel: Apollo Scientific

OR964070-25G
APOSOR964070-25GEA 2440 CZK
APOSOR964070-25G
Z-D-MEAla-OH 97%
Z-D-MEAla-OH
Vzorec: C₁₂H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00153387
CAS číslo: 68223-03-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR