Tisk…

(S)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR962590-1G OR962590-250MG OR962590-5G
APOSOR962590-1GEA 1120 CZK
APOSOR962590-1G APOSOR962590-250MG APOSOR962590-5G
(S)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid 95%
(S)-3-(tert-Butoxycarbonyl)-2,2-dimethyloxazolidine-4-carboxylic acid
Vzorec: C₁₁H₁₉NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD02682656
CAS číslo: 139009-66-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR