Tisk…

2-Bromo-1,1-dimethoxypropane 95%

Dodavatel: Apollo Scientific

OR960515-25G
APOSOR960515-25GEA 4630 CZK
APOSOR960515-25G
2-Bromo-1,1-dimethoxypropane 95%
2-Bromo-1,1-dimethoxypropane
Vzorec: C₅H₁₁BrO₂
MW: 183.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505877
CAS číslo: 33170-72-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR