Tisk…

tert-Butyl N,N-diethylcarbamate 98%

Dodavatel: Apollo Scientific

OR960322-1G OR960322-5G
APOSOR960322-1GEA 6220 CZK
APOSOR960322-1G APOSOR960322-5G
tert-Butyl N,N-diethylcarbamate 98%
tert-Butyl N,N-diethylcarbamate
Vzorec: C₉H₁₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD20929510
CAS číslo: 51170-54-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR