Tisk…

Methyl 1-methylcyclohexane-1-carboxylate 98%

Dodavatel: Apollo Scientific

OR960236-1G OR960236-5G
APOSOR960236-1GEA 5200 CZK
APOSOR960236-1G APOSOR960236-5G
Methyl 1-methylcyclohexane-1-carboxylate 98%
Methyl 1-methylcyclohexane-1-carboxylate
Vzorec: C₉H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD12025265
CAS číslo: 825-04-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR