Tisk…

3-Chloro-6-piperidin-1-ylpyridazine 98%

Dodavatel: Apollo Scientific

OR959487-10G OR959487-1G OR959487-25G
APOSOR959487-10GEA 9720 CZK
APOSOR959487-10G APOSOR959487-1G APOSOR959487-25G
3-Chloro-6-piperidin-1-ylpyridazine 98%
3-Chloro-6-piperidin-1-ylpyridazine
Vzorec: C₉H₁₂ClN₃
Storage Temperature: Ambient
MDL Number: MFCD00574572
CAS číslo: 1722-11-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR