Tisk…

N-Propyl 4-chloropicolinamide 98%

Dodavatel: Apollo Scientific

OR959443-1G OR959443-5G
APOSOR959443-1GEA 4770 CZK
APOSOR959443-1G APOSOR959443-5G
N-Propyl 4-chloropicolinamide 98%
N-Propyl 4-chloropicolinamide
Vzorec: C₉H₁₁ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD11177055
CAS číslo: 694498-91-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR