Tisk…

2,3-Dichloro-4-methoxyphenol 95%

Dodavatel: Apollo Scientific

OR957648-1G
APOSOR957648-1GEA 25820 CZK
APOSOR957648-1G
2,3-Dichloro-4-methoxyphenol 95%
2,3-Dichloro-4-methoxyphenol
Vzorec: C₇H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00155161
CAS číslo: 39542-65-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR