Tisk…

5-Hydroxy-2-isopropoxy-3-methylpyridine 95%

Dodavatel: Apollo Scientific

OR957535-1G
APOSOR957535-1GEA 17990 CZK
APOSOR957535-1G
5-Hydroxy-2-isopropoxy-3-methylpyridine 95%
5-Hydroxy-2-isopropoxy-3-methylpyridine
Vzorec: C₉H₁₃NO₂
MW: 167.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045269
CAS číslo: 1449008-14-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR