Tisk…

5-Chloro-4-hydroxy-2-methoxypyridine 97%

Dodavatel: Apollo Scientific

OR957493-1G
APOSOR957493-1GEA 10350 CZK
APOSOR957493-1G
5-Chloro-4-hydroxy-2-methoxypyridine 97%
5-Chloro-4-hydroxy-2-methoxypyridine
Vzorec: C₆H₆ClNO₂
MW: 159,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189878
CAS číslo: 1196146-71-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR