Tisk…

4-(Ethylsulphanyl)phenol 95%

Dodavatel: Apollo Scientific

OR957437-1G
APOSOR957437-1GEA 17990 CZK
APOSOR957437-1G
4-(Ethylsulphanyl)phenol 95%
4-(Ethylsulphanyl)phenol
Vzorec: C₈H₁₀OS
MW: 154.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07776119
CAS číslo: 1195-46-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR