Tisk…

2-(Ethylsulphanyl)phenol 95%

Dodavatel: Apollo Scientific

OR957435-1G
APOSOR957435-1GEA 12830 CZK
APOSOR957435-1G
2-(Ethylsulphanyl)phenol 95%
2-(Ethylsulphanyl)phenol
Vzorec: C₈H₁₀OS
MW: 154.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010815
CAS číslo: 29549-60-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR