Tisk…

5-Bromo-1-propyl-1H-pyrazole 95%

Dodavatel: Apollo Scientific

OR956656-1G
APOSOR956656-1GEA 18420 CZK
APOSOR956656-1G
5-Bromo-1-propyl-1H-pyrazole 95%
5-Bromo-1-propyl-1H-pyrazole
Vzorec: C₆H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD23134838
CAS číslo: 1427021-01-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR