Tisk…

1-tert-Butyl-3-nitrobenzene 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(tert-Butyl)-3-nitrobenzene

OR956369-5G
APOSOR956369-5GEA 1820 CZK
APOSOR956369-5G
1-tert-Butyl-3-nitrobenzene 95%
1-tert-Butyl-3-nitrobenzene
Vzorec: C₁₀H₁₃NO₂
MW: 179.22
Storage Temperature: Ambient
MDL Number: MFCD00086569
CAS číslo: 23132-52-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR