Tisk…

5-Methoxy-1,3-dimethyl-2-nitrobenzene 95%

Dodavatel: Apollo Scientific

OR956366-1G
APOSOR956366-1GEA 1370 CZK
APOSOR956366-1G
5-Methoxy-1,3-dimethyl-2-nitrobenzene 95%
5-Methoxy-1,3-dimethyl-2-nitrobenzene
Vzorec: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00506726
CAS číslo: 61019-03-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR